Assay Details
| Gene Symbol: gex-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067548.5 | NP_499949.2 | 1 - 2 | 47 | 88 | |
| GenBank mRNA | AB073209.1 | - | 1 - 2 | 69 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gex-2| Gene Symbol: gex-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067548.5 | NP_499949.2 | 1 - 2 | 47 | 88 | |
| GenBank mRNA | AB073209.1 | - | 1 - 2 | 69 | 88 | |
Set Membership: |
|