Assay Details
| Gene Symbol: ndnf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067881.5 | NP_500282.2 | 1 - 2 | 139 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ndnf-1| Gene Symbol: ndnf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067881.5 | NP_500282.2 | 1 - 2 | 139 | 94 | |
Set Membership: |
|