Assay Details
| Gene Symbol: cal-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307362.2 | NP_001294291.1 | 1 - 2 | 124 | 99 | |
| NM_068020.7 | NP_500421.2 | 2 - 3 | 325 | 99 | ||
More Information
Set Membership: |
|

cal-3
Genomic Map