Assay Details
| Gene Symbol: cal-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307362.2 | NP_001294291.1 | 5 - 6 | 458 | 76 | |
| NM_068020.7 | NP_500421.2 | 6 - 7 | 659 | 76 | ||
More Information
Set Membership: |
|

cal-3
Genomic Map