Assay Details
| Gene Symbol: lam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380214.1 | NP_001368376.1 | 1 - 2 | 91 | 70 | |
| NM_068333.6 | NP_500734.3 | 10 - 11 | 5233 | 70 | ||
More Information
Set Membership: |
|

lam-1
Genomic Map