Assay Details
| Gene Symbol: lam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068333.6 | NP_500734.3 | 5 - 6 | 1643 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lam-1| Gene Symbol: lam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068333.6 | NP_500734.3 | 5 - 6 | 1643 | 84 | |
Set Membership: |
|