Assay Details
| Gene Symbol: unc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306820.3 | NP_001293749.1 | 2 - 3 | 206 | 83 | |
| NM_001383098.1 | 1 - 2 | 126 | 83 | |||
| NM_068421.6 | NP_500822.2 | 1 - 2 | 122 | 83 | ||
| NM_068422.6 | NP_500823.1 | 1 - 2 | 124 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383098.1 transcripts.
Set Membership: |
|

unc-5
Genomic Map