Assay Details
| Gene Symbol: unc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306820.3 | NP_001293749.1 | 6 - 7 | 1053 | 159 | |
| NM_001306822.3 | NP_001293751.1 | 2 - 3 | 468 | 159 | ||
| NM_001380233.1 | NP_001368390.1 | 1 - 2 | 183 | 159 | ||
| NM_001383098.1 | 5 - 6 | 974 | 159 | |||
| NM_068421.6 | NP_500822.2 | 5 - 6 | 969 | 159 | ||
| NM_068422.6 | NP_500823.1 | 5 - 6 | 971 | 159 | ||
More Information
Set Membership: |
|

unc-5
Genomic Map