Assay Details
| Gene Symbol: aat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028211.4 | NP_001023382.1 | 1 - 2 | 97 | 91 | |
| NR_131543.1 | 1 - 2 | 89 | 91 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aat-4| Gene Symbol: aat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028211.4 | NP_001023382.1 | 1 - 2 | 97 | 91 | |
| NR_131543.1 | 1 - 2 | 89 | 91 | |||
Set Membership: |
|