Assay Details
| Gene Symbol: aat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028211.4 | NP_001023382.1 | 4 - 5 | 458 | 113 | |
| NR_131543.1 | 4 - 5 | 467 | 113 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aat-4| Gene Symbol: aat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028211.4 | NP_001023382.1 | 4 - 5 | 458 | 113 | |
| NR_131543.1 | 4 - 5 | 467 | 113 | |||
Set Membership: |
|