Assay Details
| Gene Symbol: smg-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380273.1 | NP_001368064.1 | 1 - 2 | 263 | 130 | |
| NM_068573.9 | NP_500974.2 | 1 - 2 | 267 | 130 | ||
| GenBank mRNA | EF472000.1 | - | 1 - 2 | 281 | 130 | |
More Information
Set Membership: |
|

smg-3
Genomic Map