Assay Details
| Gene Symbol: smg-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380273.1 | NP_001368064.1 | 9 - 10 | 2715 | 137 | |
| NM_068573.9 | NP_500974.2 | 9 - 10 | 2719 | 137 | ||
| GenBank mRNA | EF472000.1 | - | 9 - 10 | 2733 | 137 | |
More Information
Set Membership: |
|

smg-3
Genomic Map