Assay Details
| Gene Symbol: lpr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069830.5 | NP_502231.1 | 18 - 19 | 2612 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lpr-2| Gene Symbol: lpr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069830.5 | NP_502231.1 | 18 - 19 | 2612 | 70 | |
Set Membership: |
|