Assay Details
| Gene Symbol: efa-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129346.3 | NP_001122818.1 | 1 - 2 | 162 | 59 | |
| NM_070015.6 | NP_502416.1 | 2 - 3 | 214 | 59 | ||
| NM_070016.6 | NP_502417.1 | 2 - 3 | 220 | 59 | ||
| GenBank mRNA | EF473217.1 | - | 1 - 2 | 157 | 59 | |
| EF473218.1 | - | 1 - 2 | 157 | 59 | ||
More Information
Set Membership: |
|

efa-6
Genomic Map