Assay Details
| Gene Symbol: efa-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070015.6 | NP_502416.1 | 3 - 4 | 353 | 60 | |
| GenBank mRNA | EF473217.1 | - | 2 - 3 | 296 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

efa-6| Gene Symbol: efa-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070015.6 | NP_502416.1 | 3 - 4 | 353 | 60 | |
| GenBank mRNA | EF473217.1 | - | 2 - 3 | 296 | 60 | |
Set Membership: |
|