Assay Details
| Gene Symbol: efa-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268723.3 | NP_001255652.1 | 1 - 2 | 143 | 85 | |
| NM_070015.6 | NP_502416.1 | 5 - 6 | 752 | 85 | ||
| NM_070016.6 | NP_502417.1 | 5 - 6 | 752 | 85 | ||
| GenBank mRNA | EF473217.1 | - | 4 - 5 | 695 | 85 | |
More Information
Set Membership: |
|

efa-6
Genomic Map