Assay Details
| Gene Symbol: efa-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129346.3 | NP_001122818.1 | 8 - 9 | 2121 | 67 | |
| NM_001268723.3 | NP_001255652.1 | 6 - 7 | 1723 | 67 | ||
| NM_070015.6 | NP_502416.1 | 10 - 11 | 2332 | 67 | ||
| NM_070016.6 | NP_502417.1 | 10 - 11 | 2332 | 67 | ||
| GenBank mRNA | EF473217.1 | - | 9 - 10 | 2275 | 67 | |
| EF473218.1 | - | 8 - 9 | 2116 | 67 | ||
More Information
Set Membership: |
|

efa-6
Genomic Map