Assay Details
| Gene Symbol: rpl-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047495.6 | NP_001040960.1 | 1 - 2 | 59 | 88 | |
| NM_070141.9 | NP_502542.1 | 3 - 4 | 495 | 88 | ||
| NR_002385.1 | 3 - 4 | 523 | 88 | |||
More Information
Set Membership: |
|

rpl-12
Genomic Map