Assay Details
| Gene Symbol: drh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380283.2 | NP_001368373.1 | 8 - 9 | 1094 | 100 | |
| NM_068617.8 | NP_501018.1 | 13 - 14 | 1873 | 100 | ||
| GenBank mRNA | AF480439.1 | - | 13 - 14 | 1868 | 100 | |
More Information
Set Membership: |
|

drh-1
Genomic Map