Assay Details
| Gene Symbol: drh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380283.2 | NP_001368373.1 | 12 - 13 | 2053 | 91 | |
| NM_068617.8 | NP_501018.1 | 17 - 18 | 2832 | 91 | ||
| GenBank mRNA | AF480439.1 | - | 17 - 18 | 2827 | 91 | |
More Information
Set Membership: |
|

drh-1
Genomic Map