Assay Details
| Gene Symbol: drh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068617.8 | NP_501018.1 | 1 - 2 | 95 | 107 | |
| GenBank mRNA | AF480439.1 | - | 1 - 2 | 90 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

drh-1| Gene Symbol: drh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068617.8 | NP_501018.1 | 1 - 2 | 95 | 107 | |
| GenBank mRNA | AF480439.1 | - | 1 - 2 | 90 | 107 | |
Set Membership: |
|