Assay Details
| Gene Symbol: drh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068617.8 | NP_501018.1 | 2 - 3 | 267 | 109 | |
| GenBank mRNA | AF480439.1 | - | 2 - 3 | 262 | 109 | |
More Information
Set Membership: |
|

drh-1
Genomic Map