Assay Details
| Gene Symbol: tre-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306844.3 | NP_001293773.1 | 4 - 5 | 420 | 92 | |
| NM_001380293.1 | NP_001368400.1 | 3 - 4 | 279 | 92 | ||
| NM_068657.7 | NP_501058.2 | 6 - 7 | 678 | 92 | ||
| GenBank mRNA | AJ512334.1 | - | 5 - 6 | 655 | 92 | |
More Information
Set Membership: |
|

tre-2
Genomic Map