Assay Details
| Gene Symbol: tre-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306844.3 | NP_001293773.1 | 6 - 7 | 683 | 88 | |
| NM_001380293.1 | NP_001368400.1 | 5 - 6 | 542 | 88 | ||
| NM_068657.7 | NP_501058.2 | 8 - 9 | 941 | 88 | ||
| GenBank mRNA | AJ512334.1 | - | 7 - 8 | 918 | 88 | |
More Information
Set Membership: |
|

tre-2
Genomic Map