Assay Details
| Gene Symbol: dmd-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070775.5 | NP_503176.2 | 2 - 3 | 328 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dmd-8| Gene Symbol: dmd-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070775.5 | NP_503176.2 | 2 - 3 | 328 | 56 | |
Set Membership: |
|