Assay Details
| Gene Symbol: tat-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071457.2 | NP_503858.1 | 7 - 8 | 2877 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tat-6| Gene Symbol: tat-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071457.2 | NP_503858.1 | 7 - 8 | 2877 | 108 | |
Set Membership: |
|