Assay Details
| Gene Symbol: sos-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373020.3 | NP_001360576.1 | 10 | 2710 | 65 | |
| NM_071834.5 | NP_504235.2 | 10 - 11 | 2722 | 65 | ||
More Information
Set Membership: |
|

sos-1
Genomic Map