Assay Details
| Gene Symbol: sos-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373020.3 | NP_001360576.1 | 4 | 599 | 73 | |
| NM_071834.5 | NP_504235.2 | 4 - 5 | 611 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sos-1| Gene Symbol: sos-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373020.3 | NP_001360576.1 | 4 | 599 | 73 | |
| NM_071834.5 | NP_504235.2 | 4 - 5 | 611 | 73 | ||
Set Membership: |
|