Assay Details
| Gene Symbol: ncx-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071940.7 | NP_504341.1 | 8 - 9 | 1546 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ncx-8| Gene Symbol: ncx-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071940.7 | NP_504341.1 | 8 - 9 | 1546 | 150 | |
Set Membership: |
|