Assay Details
| Gene Symbol: lgc-39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072420.5 | NP_504821.3 | 4 - 5 | 877 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lgc-39| Gene Symbol: lgc-39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072420.5 | NP_504821.3 | 4 - 5 | 877 | 111 | |
Set Membership: |
|