Assay Details
| Gene Symbol: lmd-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373062.3 | NP_001360511.1 | 12 - 13 | 1835 | 97 | |
| NM_001373063.2 | NP_001360537.1 | 11 - 12 | 1796 | 97 | ||
| NM_001373064.1 | NP_001360538.1 | 11 - 12 | 1802 | 97 | ||
| NM_072772.4 | NP_505173.2 | 12 - 13 | 1859 | 97 | ||
| NM_072773.7 | NP_505174.3 | 12 - 13 | 1865 | 97 | ||
More Information
Set Membership: |
|

lmd-3
Genomic Map