Assay Details
| Gene Symbol: selt-1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073340.3 | NP_505741.2 | 3 - 4 | 376 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

selt-1.2| Gene Symbol: selt-1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073340.3 | NP_505741.2 | 3 - 4 | 376 | 153 | |
Set Membership: |
|