Assay Details
| Gene Symbol: scb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269371.3 | NP_001256300.1 | 1 - 2 | 68 | 106 | |
| NM_001383400.1 | 8 - 9 | 2415 | 106 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scb-1| Gene Symbol: scb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269371.3 | NP_001256300.1 | 1 - 2 | 68 | 106 | |
| NM_001383400.1 | 8 - 9 | 2415 | 106 | |||
Set Membership: |
|