Assay Details
| Gene Symbol: scb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269371.3 | NP_001256300.1 | 2 - 3 | 215 | 69 | |
| NM_001383400.1 | 9 - 10 | 2562 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scb-1| Gene Symbol: scb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269371.3 | NP_001256300.1 | 2 - 3 | 215 | 69 | |
| NM_001383400.1 | 9 - 10 | 2562 | 69 | |||
Set Membership: |
|