Assay Details
| Gene Symbol: kmo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136430.1 | NP_001129902.1 | 1 - 2 | 291 | 84 | |
| NM_001373128.1 | NP_001359579.1 | 1 - 2 | 229 | 84 | ||
More Information
Set Membership: |
|

kmo-2
Genomic Map