Assay Details
| Gene Symbol: rad-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129527.3 | NP_001122999.1 | 11 - 12 | 3516 | 156 | |
| NM_073669.8 | NP_506070.1 | 10 - 11 | 3422 | 156 | ||
| GenBank mRNA | Z75312.1 | - | 10 - 11 | 3452 | 156 | |
More Information
Set Membership: |
|

rad-50
Genomic Map