Assay Details
| Gene Symbol: rad-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129527.3 | NP_001122999.1 | 12 - 13 | 3822 | 83 | |
| NM_073669.8 | NP_506070.1 | 11 - 12 | 3728 | 83 | ||
| GenBank mRNA | Z75312.1 | - | 11 - 12 | 3758 | 83 | |
More Information
Set Membership: |
|

rad-50
Genomic Map