Assay Details
| Gene Symbol: rad-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129527.3 | NP_001122999.1 | 8 - 9 | 1163 | 101 | |
| NM_073669.8 | NP_506070.1 | 7 - 8 | 1069 | 101 | ||
| GenBank mRNA | Z75312.1 | - | 7 - 8 | 1099 | 101 | |
More Information
Set Membership: |
|

rad-50
Genomic Map