Assay Details
| Gene Symbol: gcy-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073696.3 | NP_506097.3 | 17 - 18 | 3037 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gcy-13| Gene Symbol: gcy-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073696.3 | NP_506097.3 | 17 - 18 | 3037 | 140 | |
Set Membership: |
|