Assay Details
| Gene Symbol: set-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269683.3 | NP_001256612.1 | 8 - 9 | 3271 | 121 | |
| NM_001269684.3 | NP_001256613.1 | 7 - 8 | 2583 | 121 | ||
| NM_001269685.3 | NP_001256614.1 | 7 - 8 | 2583 | 121 | ||
| NM_001276960.4 | NP_001263889.1 | 8 - 9 | 3267 | 121 | ||
More Information
Set Membership: |
|

set-5
Genomic Map