Assay Details
| Gene Symbol: set-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269683.3 | NP_001256612.1 | 12 - 13 | 3814 | 89 | |
| NM_001269684.3 | NP_001256613.1 | 11 - 12 | 3135 | 89 | ||
| NM_001269685.3 | NP_001256614.1 | 11 - 12 | 3126 | 89 | ||
| NM_001276960.4 | NP_001263889.1 | 12 - 13 | 3810 | 89 | ||
More Information
Set Membership: |
|

set-5
Genomic Map