Assay Details
| Gene Symbol: set-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269683.3 | NP_001256612.1 | 15 - 16 | 4250 | 85 | |
| NM_001269684.3 | NP_001256613.1 | 14 - 15 | 3571 | 85 | ||
| NM_001269685.3 | NP_001256614.1 | 14 - 15 | 3562 | 85 | ||
| NM_001276960.4 | NP_001263889.1 | 14 - 15 | 4132 | 85 | ||
More Information
Set Membership: |
|

set-5
Genomic Map