Assay Details
| Gene Symbol: set-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269683.3 | NP_001256612.1 | 17 - 18 | 4618 | 73 | |
| NM_001269684.3 | NP_001256613.1 | 16 - 17 | 3939 | 73 | ||
| NM_001269685.3 | NP_001256614.1 | 16 - 17 | 3930 | 73 | ||
| NM_001276960.4 | NP_001263889.1 | 16 - 17 | 4500 | 73 | ||
More Information
Set Membership: |
|

set-5
Genomic Map