Assay Details
| Gene Symbol: set-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269683.3 | NP_001256612.1 | 1 - 2 | 571 | 95 | |
| NM_001276960.4 | NP_001263889.1 | 1 - 2 | 567 | 95 | ||
More Information
Set Membership: |
|

set-5
Genomic Map