Assay Details
| Gene Symbol: mpst-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269701.3 | NP_001256630.1 | 5 - 6 | 920 | 100 | |
| NM_001269702.4 | NP_001256631.1 | 6 - 7 | 830 | 100 | ||
| NM_001269703.2 | NP_001256632.1 | 4 - 5 | 645 | 100 | ||
More Information
Set Membership: |
|

mpst-3
Genomic Map