Assay Details
| Gene Symbol: cdr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074585.9 | NP_506986.1 | 3 - 4 | 420 | 76 | |
| GenBank mRNA | AF301606.1 | - | 3 - 4 | 368 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdr-1| Gene Symbol: cdr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074585.9 | NP_506986.1 | 3 - 4 | 420 | 76 | |
| GenBank mRNA | AF301606.1 | - | 3 - 4 | 368 | 76 | |
Set Membership: |
|