Assay Details
| Gene Symbol: cwc-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074623.6 | NP_507024.1 | 1 - 2 | 177 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cwc-15| Gene Symbol: cwc-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074623.6 | NP_507024.1 | 1 - 2 | 177 | 64 | |
Set Membership: |
|