Assay Details
| Gene Symbol: cth-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276977.3 | NP_001263906.1 | 1 - 2 | 602 | 64 | |
| NM_074652.5 | NP_507053.1 | 2 - 3 | 766 | 64 | ||
More Information
Set Membership: |
|

cth-1
Genomic Map