Assay Details
| Gene Symbol: efl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074888.6 | NP_507289.1 | 3 - 4 | 388 | 99 | |
| GenBank mRNA | AY028166.1 | - | 3 - 4 | 395 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

efl-1| Gene Symbol: efl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074888.6 | NP_507289.1 | 3 - 4 | 388 | 99 | |
| GenBank mRNA | AY028166.1 | - | 3 - 4 | 395 | 99 | |
Set Membership: |
|