Assay Details
| Gene Symbol: efl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074888.6 | NP_507289.1 | 4 - 5 | 545 | 137 | |
| GenBank mRNA | AY028166.1 | - | 4 - 5 | 552 | 137 | |
More Information
Set Membership: |
|

efl-1
Genomic Map